MilliporeSigma
  • Loss of ISWI Function in Drosophila Nuclear Bodies Drives Cytoplasmic Redistribution of Drosophila TDP-43.
Loss of ISWI Function in Drosophila Nuclear Bodies Drives Cytoplasmic Redistribution of Drosophila TDP-43.
International journal of molecular sciences (2018-04-05)
Luca Lo Piccolo, Rosa Bonaccorso, Andrea Attardi, Lorenzo Li Greci, Giulia Romano, Martina Sollazzo, Giorgio Giurato, Antonia Maria Rita Ingrassia, Fabian Feiguin, Davide F V Corona, Maria Cristina Onorati
ABSTRACT
Over the past decade, evidence has identified a link between protein aggregation, RNA biology, and a subset of degenerative diseases. An important feature of these disorders is the cytoplasmic or nuclear aggregation of RNA-binding proteins (RBPs). Redistribution of RBPs, such as the human TAR DNA-binding 43 protein (TDP-43) from the nucleus to cytoplasmic inclusions is a pathological feature of several diseases. Indeed, sporadic and familial forms of amyotrophic lateral sclerosis (ALS) and fronto-temporal lobar degeneration share as hallmarks ubiquitin-positive inclusions. Recently, the wide spectrum of neurodegenerative diseases characterized by RBPs functions' alteration and loss was collectively named proteinopathies. Here, we show that TBPH (TAR DNA-binding protein-43 homolog), the Drosophila ortholog of human TDP-43 TAR DNA-binding protein-43, interacts with the arcRNA hsrω and with hsrω-associated hnRNPs. Additionally, we found that the loss of the omega speckles remodeler ISWI (Imitation SWI) changes the TBPH sub-cellular localization to drive a TBPH cytoplasmic accumulation. Our results, hence, identify TBPH as a new component of omega speckles and highlight a role of chromatin remodelers in hnRNPs nuclear compartmentalization.
Sigma-Aldrich
Tetrabutylphosphonium hydroxide solution, 40 wt. % in H2O
Pricing and availability is not currently available.