MilliporeSigma
  • Synthesis and antimicrobial activity of cholic acid hydrazone analogues.
Synthesis and antimicrobial activity of cholic acid hydrazone analogues.
European journal of medicinal chemistry (2010-02-26)
Anas J M Rasras, Taleb H Al-Tel, Amal F Al-Aboudi, Raed A Al-Qawasmeh
ABSTRACT
Synthesis and antimicrobial activity of cholic acid analogues 4a-t are reported. The synthesis of 4a-t was accomplished from ethylcholate 2. The hydrazone moiety was introduced via coupling of the cholic acid hydrazide (3) with appropriately functionalized aldehyde utilizing acetic acid as a catalyst. Quiet of interest in relation to the synthesized hydrazones is the formation of two rotamers s-cis.E and s-trans.E. Most compounds showed stronger antimicrobial activity against Gram-positive bacteria than Cefaclor and Cefixime. Compounds 4d, 4i and 4j indicated 15-fold stronger antimicrobial activities against Enterobacter faecalis compared to Cefaclor and Cefixime. Some of the synthesized compounds (e.g. 4a, 4c, 4d, 4i, and 4l) reflected two-folds less activity against Escherichia coli relative to Cefixime.
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
5-Methoxyindole-3-carboxaldehyde, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cefaclor
Pricing and availability is not currently available.