MilliporeSigma
  • Mitochondrial Complex I activity signals antioxidant response through ERK5.
Mitochondrial Complex I activity signals antioxidant response through ERK5.
Scientific reports (2018-05-11)
Abrar Ul Haq Khan, Nerea Allende-Vega, Delphine Gitenay, Johan Garaude, Dang-Nghiem Vo, Sana Belkhala, Sabine Gerbal-Chaloin, Claire Gondeau, Martine Daujat-Chavanieu, Cécile Delettre, Stefania Orecchioni, Giovanna Talarico, Francesco Bertolini, Alberto Anel, José M Cuezva, Jose A Enriquez, Guillaume Cartron, Charles-Henri Lecellier, Javier Hernandez, Martin Villalba
ABSTRACT
Oxidative phosphorylation (OXPHOS) generates ROS as a byproduct of mitochondrial complex I activity. ROS-detoxifying enzymes are made available through the activation of their antioxidant response elements (ARE) in their gene promoters. NRF2 binds to AREs and induces this anti-oxidant response. We show that cells from multiple origins performing OXPHOS induced NRF2 expression and its transcriptional activity. The NRF2 promoter contains MEF2 binding sites and the MAPK ERK5 induced MEF2-dependent NRF2 expression. Blocking OXPHOS in a mouse model decreased Erk5 and Nrf2 expression. Furthermore, fibroblasts derived from patients with mitochondrial disorders also showed low expression of ERK5 and NRF2 mRNAs. Notably, in cells lacking functional mitochondrial complex I activity OXPHOS did not induce ERK5 expression and failed to generate this anti-oxidant response. Complex I activity induces ERK5 expression through fumarate accumulation. Eukaryotic cells have evolved a genetic program to prevent oxidative stress directly linked to OXPHOS and not requiring ROS.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK7