MilliporeSigma
IGFBP-6 five years on; not so 'forgotten'?
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2005-05-26)
Leon A Bach
ABSTRACT
Insulin-like growth factor binding protein (IGFBP)-6 is unique among IGFBPs for its IGF-II binding specificity. IGFBP-6 inhibits growth of a number of IGF-II-dependent cancers, including rhabdomyosarcoma, neuroblastoma and colon cancer. Although the major action of IGFBP-6 appears to be inhibition of IGF-II actions, a number of studies suggest that it may also have IGF-independent actions. Gene array studies show regulation of IGFBP-6 in many circumstances that are consistent with antiproliferative actions. However, other studies show the opposite, so that IGFBP-6 may be acting as a counter-regulator in these situations or it may have other as yet undetermined actions. Both the N-terminal and C-terminal domains of IGFBP-6 contribute to high affinity IGF binding, and the C-terminal domain appears to confer its IGF-II specificity. The three-dimensional structure of the C-domain of IGFBP-6 contains a thyroglobulin type 1 fold, and the IGF-II binding site is located in the proximal half of this domain adjacent to the glycosaminoglycan binding site. Future studies are needed to further delineate the putative IGF-independent actions of IGFBP-6 and to build on the structural information to enhance our understanding of this IGFBP. This is particularly significant since IGFBP-6 provides an attractive basis for therapy of IGF-II-dependent tumors.
Sigma-Aldrich
IGF-Binding Protein 6 human, recombinant, expressed in (BTI-Tn-5B1-4) High-5 Insect Cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture