MilliporeSigma
Chemokines and the tissue-specific migration of lymphocytes.
Immunity (2002-02-05)
Eric J Kunkel, Eugene C Butcher
ABSTRACT
Tissue-selective trafficking of memory and effector T and B lymphocytes is mediated by unique combinations of adhesion molecules and chemokines. The discovery of several related epithelial-expressed chemokines (TECK/CCL25 in small intestine, CTACK/CCL27 in skin, and MEC/CCL28 in diverse mucosal sites) now highlights an important role for epithelial cells in controlling homeostatic lymphocyte trafficking, including the localization of cutaneous and intestinal memory T cells, and of IgA plasma cells. Constitutively expressed epithelial chemokines may help determine the character of local immune responses and contribute to the systemic organization of the immune system.
Sigma-Aldrich
M-CSF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
M-CSF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
MEC (CCL28) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture