MilliporeSigma
  • Mammalian neurotrophin-4: structure, chromosomal localization, tissue distribution, and receptor specificity.
Mammalian neurotrophin-4: structure, chromosomal localization, tissue distribution, and receptor specificity.
Proceedings of the National Academy of Sciences of the United States of America (1992-04-01)
N Y Ip, C F Ibáñez, S H Nye, J McClain, P F Jones, D R Gies, L Belluscio, M M Le Beau, R Espinosa, S P Squinto
ABSTRACT
Nerve growth factor, brain-derived neurotrophic factor, and neurotrophin-3 (NT-3) are the three members of the neurotrophin family known to exist in mammals. Recently, a fourth neurotrophin (designated neurotrophin-4 or NT-4), which shares all of the features found in the mammalian neurotrophins, has been identified in Xenopus and viper. We used sequences specific to the Xenopus/viper NT-4 to isolate a neurotrophin from both human and rat genomic DNA that appears to represent the mammalian counterpart of Xenopus/viper NT-4. Human NT-4 as well as a human NT-4 pseudogene colocalize to chromosome 19 band q13.3. Mammalian NT-4 has many unusual features compared to the previously identified neurotrophins and is less conserved evolutionarily than the other neurotrophins. However, mammalian NT-4 displays bioactivity and trk receptor specificity similar to that of Xenopus NT-4.
Sigma-Aldrich
NT-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture