MilliporeSigma
  • Endoscopy, histology and electron microscopy analysis of foetal membranes in pregnant South American plains vizcacha reveal unusual excrescences on the yolk sac.
Endoscopy, histology and electron microscopy analysis of foetal membranes in pregnant South American plains vizcacha reveal unusual excrescences on the yolk sac.
Journal of molecular histology (2018-02-23)
Mariela Giacchino, Pablo I F Inserra, Fernando D Lange, María C Gariboldi, Sergio R Ferraris, Alfredo D Vitullo
ABSTRACT
The South American hystricognathe Lagostomus maximus is a fossorial rodent whose females show unique reproductive characteristics. They have a 155-day long gestation, show massive polyovulation and a selective process of embryonic resorption in the first half of gestation. In order to explore and perform an in-situ characterization of the reproductive tract, we visualized internal structures through ultrasonography and video-endoscopy in pregnant and non-pregnant females. We describe the finding of protruding structures that lie on the yolk sac and their histological and ultrastructural characterization. The placenta was covered with whitish, small pearl-shaped structures. These structures were also seen on the extra-embryonic space, being the amnion and the umbilical cord free of them. Pearl-shaped structures were composed with loose connective tissue, lacked blood vessels, and showed collagen fibers organized in a spiral form. They were anchored by pedicles to the villous surface of the extraembryonic membrane. We discuss the biological and evolutionary meaning of the pearl-shaped structures that relate L. maximus to the African origin of the South American hystricognathe fauna.
Product Description
Ethanol solution, NIST® SRM® 2899a, nominal 25% by mass