MilliporeSigma
  • Heme oxygenase-1 protects liver against ischemia/reperfusion injury via phosphoglycerate mutase family member 5-mediated mitochondrial quality control.
Heme oxygenase-1 protects liver against ischemia/reperfusion injury via phosphoglycerate mutase family member 5-mediated mitochondrial quality control.
Life sciences (2018-03-11)
Jeong-Min Hong, Sun-Mee Lee
ABSTRACT
Heme oxygenase-1 (HO-1), an endogenous cytoprotective enzyme, is reported that can be localized in mitochondria under stress, contributing to preserve mitochondrial function. Mitochondrial quality control (QC) is essential to cellular health and recovery linked with redox homeostasis. Recent studies reported that phosphoglycerate mutase family member (PGAM) 5, a mitochondria-resident phosphatase, plays critical role in mitochondrial homeostasis. Therefore, we aim to investigate cytoprotective mechanisms of HO-1 in I/R-induced hepatic injury focusing on mitochondrial QC associated with PGAM5 signaling. Mice were subjected to 60 min of hepatic ischemia followed by 6 h reperfusion and were pretreated twice with hemin (HO-1 inducer, 30 mg/kg) or zinc protoporphyrin (ZnPP; HO-1 inhibitor, 10 mg/kg) 16 and 3 h before ischemia. I/R increased hepatic and mitochondrial HO activity, which was augmented by hemin. I/R-induced hepatocellular and mitochondrial damages were attenuated by hemin and augmented by ZnPP. Meanwhile, I/R increased mitochondrial biogenesis, as evidenced by increased mitochondrial DNA contents and mitochondrial transcription factor A protein expression. Hemin augmented these results. I/R impaired mitophagy, as indicated by decreases in Parkin protein expression and the number of mitophagic vacuoles. These changes were attenuated by hemin. Hemin attenuated the I/R-induced increase in mitochondrial fission-related protein, dynamin-related protein 1, and the decrease in PGAM5 protein expression. Furthermore, PGAM5 siRNA abolished the effect of HO-1 on mitochondrial QC in HepG2 cells subjected to hypoxia/reoxygenation. Our findings suggest that HO-1 protects against I/R-induced hepatic injury via regulation of mitochondrial QC by PGAM5 signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human PGAM5