MilliporeSigma
  • Synthesis of High Relaxivity Gadolinium AAZTA Tetramers as Building Blocks for Bioconjugation.
Synthesis of High Relaxivity Gadolinium AAZTA Tetramers as Building Blocks for Bioconjugation.
Bioconjugate chemistry (2018-02-23)
Martina Tripepi, Federico Capuana, Eliana Gianolio, Flávio Vinicius Crizóstomo Kock, Amerigo Pagoto, Rachele Stefania, Giuseppe Digilio, Silvio Aime
ABSTRACT
Molecular imaging requires the specific accumulation of contrast agents at the target. To exploit the superb resolution of MRI for applications in molecular imaging, gadolinium chelates, as the MRI contrast agents (CA), have to be conjugated to a specific vector able to recognize the epitope of interest. Several Gd(III)-chelates can be chemically linked to the same binding vector in order to deliver multiple copies of the CA (multimers) in a single targeting event thus increasing the sensitivity of the molecular probe. Herein three novel bifunctional agents, carrying one functional group for the bioconjugation to targeting vectors and four Gd(III)-AAZTA chelate functions for MRI contrast enhancement (AAZTA = 6-amino-6-methylperhydro-1,4-diazepinetetraacetic acid), are reported. The relaxivity in the tetrameric derivatives is 16.4 ± 0.2 mMGd-1 s-1 at 21.5 MHz and 25 °C, being 2.4-fold higher than that of parent, monomeric Gd(III)-AAZTA. These compounds can be used as versatile building blocks to insert preformed, high relaxivity, and high density Gd-centers to biological targeting vectors. As an example, we describe the use of these bifunctional Gd(III)-chelates to label a fibrin-targeting peptide.
Sigma-Aldrich
Fmoc-Lys(Fmoc)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
2-(2-Aminoethoxy)ethylamine, AldrichCPR
Sigma-Aldrich
HBTU, ≥98.0% (T)