MilliporeSigma
  • Self-Assembled Biomolecular 1D Nanostructures for Aqueous Sodium-Ion Battery.
Self-Assembled Biomolecular 1D Nanostructures for Aqueous Sodium-Ion Battery.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2018-03-30)
Huiwu Long, Wen Zeng, Hua Wang, Mengmeng Qian, Yanhong Liang, Zhongchang Wang
ABSTRACT
Aqueous sodium-ion battery of low cost, inherent safety, and environmental benignity holds substantial promise for new-generation energy storage applications. However, the narrow potential window of water and the enlarged ionic radius because of hydration restrict the selection of electrode materials used in the aqueous electrolyte. Here, inspired by the efficient redox reaction of biomolecules during cellular energy metabolism, a proof of concept is proposed that the redox-active biomolecule alizarin can act as a novel electrode material for the aqueous sodium-ion battery. It is demonstrated that the specific capacity of the self-assembled alizarin nanowires can reach as high as 233.1 mA h g-1, surpassing the majority of anodes ever utilized in the aqueous sodium-ion batteries. Paired with biocompatible and biodegradable polypyrrole, this full battery system shows excellent sodium storage ability and flexibility, indicating its potential applications in wearable electronics and biointegrated devices. It is also shown that the electrochemical properties of electrodes can be tailored by manipulating naturally occurring 9,10-anthroquinones with various substituent groups, which broadens application prospect of biomolecules in aqueous sodium-ion batteries.
Sigma-Aldrich
Alizarin, Dye content 97 %