MilliporeSigma
  • CD9-positive exosomes from cancer-associated fibroblasts stimulate the migration ability of scirrhous-type gastric cancer cells.
CD9-positive exosomes from cancer-associated fibroblasts stimulate the migration ability of scirrhous-type gastric cancer cells.
British journal of cancer (2018-02-14)
Yuichiro Miki, Masakazu Yashiro, Tomohisa Okuno, Kishu Kitayama, Go Masuda, Kosei Hirakawa, Masaichi Ohira
ABSTRACT
This corrects the article DOI: 10.1038/bjc.2017.85.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD9