MilliporeSigma
  • The function of Notch1 intracellular domain in the differentiation of gastric cancer.
The function of Notch1 intracellular domain in the differentiation of gastric cancer.
Oncology letters (2018-04-05)
Sunkuan Hu, Qiuxiang Chen, Tiesu Lin, Wandong Hong, Wenzhi Wu, Ming Wu, Xiaojing Du, Rong Jin
ABSTRACT
Due to the complex function of the Notch signal pathway in gastric cancer (GC), the association between Notch homolog 1 (Notch1) intracellular domain (NICD) and differentiation of GC remains unknown. The present study aimed to investigate the potential association between NICD and GC differentiation, and demonstrated that poorly differentiated GC expressed increased NICD levels compared with well differentiated GC. A γ-secretase inhibitor inhibited the growth of AGS cells through downregulating NICD level. Additional data suggested that a COX-2 inhibitor caused a marked reduction of NICD level in comparison with a control group treated with dimethyl sulfoxide. Combined administration of γ-secretase and COX-2 inhibitor produced a marked inhibition of growth in AGS cells, which suggests that patients with poorly differentiated GC may benefit from the blockage of NICD, which potentially serves a role in GC differentiation.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Notch1 Antibody, Intracellular, serum, from rabbit