MilliporeSigma
  • 2-Carboxyethylgermanium sesquioxide, a synthetic organogermanium compound, as an inducer of contrasuppressor T cells.
2-Carboxyethylgermanium sesquioxide, a synthetic organogermanium compound, as an inducer of contrasuppressor T cells.
Experientia (1996-02-15)
K Ikemoto, M Kobayashi, T Fukumoto, M Morimatsu, R B Pollard, F Suzuki
ABSTRACT
2-Carboxyethylgermanium sesquioxide (Ge-132), a synthesized organogermanium compound with immunomodulating activities, was shown to be an inducer of anti-suppressor T cells in normal mice. The suppressor cell activity of T6S cells, a clone of burn-induced CD8+ IL-4-producing suppressor T cells, was clearly inhibited when a mixed lymphocyte-tumor cell reaction of the clone was conducted with splenic mononuclear cells from mice treated orally with a 100 mg/kg dose of Ge-132. The activity of anit-suppressor cells was demonstrated in spleens of mice 2 days after treatment with Ge-132 and reached its peak on day 3. The anti-suppressor cells induced by the compound were of a contrasuppressor T cell-linage, because they were characterized as CD4+ CD28+ TCRalpha/beta+ Vicia villosa lectin-adherent T cells. These cells produced IFN-gamma but did not produce IL-2, IL-4, IL-6 or IL-10 in their culture fluids. CD4+ anti-suppressor T cells induced by Ge-132 may be different from other subsets of CD4+ T cells because Th1 and Th2 cells generated in our laboratory did not adhere to Vicia villosa lectin-coated petri dishes, and each produced specific cytokines. Th1 cells produced IFN-gamma and IL-2 while Th2 cells produce IL-4 and IL-10 in vitro. These results suggest that Ge-132 may be useful as an inducer of contrasuppressor T cells in immunocompromised individuals bearing suppressor T cells. To eliminate suppressor T cells from immunocompromised hosts may result in improved resistance from various opportunistic infections.