MilliporeSigma
PD-L1 expression in medulloblastoma: an evaluation by subgroup.
Oncotarget (2018-05-04)
Allison M Martin, Christopher J Nirschl, Magda J Polanczyk, W Robert Bell, Thomas R Nirschl, Sarah Harris-Bookman, Jillian Phallen, Jessica Hicks, Daniel Martinez, Aleksandra Ogurtsova, Haiying Xu, Lisa M Sullivan, Alan K Meeker, Eric H Raabe, Kenneth J Cohen, Charles G Eberhart, Peter C Burger, Mariarita Santi, Janis M Taube, Drew M Pardoll, Charles G Drake, Michael Lim
ABSTRACT
This study evaluated the expression of PD-L1 and markers of immune mediated resistance in human medulloblastoma (MB), the most common malignant pediatric brain tumor. Overall levels of PD-L1 in human MB were low; however, some cases demonstrated robust focal expression associated with increased immune infiltrates. The case with highest PD-L1 expression was a sonic hedgehog (SHH) MB. In cell lines, SHH MB, which are low-MYC expressing, demonstrated both constitutive and inducible expression of PD-L1 while those in Group 3/4 that expressed high levels of MYC had only inducible expression. In vitro, IFN-γ robustly stimulated the expression of PD-L1 in all cell lines while radiation induced variable expression. Forced high MYC expression did not significantly alter PD-L1. Human MB tumor samples were evaluated for expression of PD-L1 and immune cell markers in relation to molecular subgroup assignment. PD-L1 expression was functionally analyzed under conditions of interferon gamma (IFN-γ), radiation, and MYC overexpression. MB expresses low levels of PD-L1 facilitating immune escape. Importantly, TH1 cytokine stimulation appears to be the most potent inducer of PD-L1 expression in vitro suggesting that an inflamed tumor microenvironment is necessary for PD-1 pathway activation in this tumor.
Sigma-Aldrich
Anti-p53 antibody, Mouse monoclonal, clone BP53-12, ascites fluid