MilliporeSigma
  • Neuroprotective Effects of CGP3466B on Apoptosis Are Modulated by Protein-L-isoaspartate (D-aspartate) O-methyltransferase/Mst1 Pathways after Traumatic Brain Injury in Rats.
Neuroprotective Effects of CGP3466B on Apoptosis Are Modulated by Protein-L-isoaspartate (D-aspartate) O-methyltransferase/Mst1 Pathways after Traumatic Brain Injury in Rats.
Scientific reports (2017-08-25)
Feng Liang, Ligen Shi, Jingwei Zheng, Sheng Chen, Yangxin Wang, Jianmin Zhang
ABSTRACT
Neuronal apoptosis chiefly contributes to the cell loss following traumatic brain injury (TBI). CGP3466B is a compound related to the anti-Parkinsonism drug R-(-)-deprenyl. Previous studies have illuminated anti-apoptosis effects of CGP3466B in different cell lines, but the underlying mechanisms have not been fully elucidated. Mammalian sterile 20 (STE20)-like kinase1 (Mst1) is a core component of the Hippo signaling pathway. Protein-L-isoaspartate (D-aspartate) O-methyltransferase (PCMT1) is an enzyme that repairs damaged L-isoaspartyl residues in proteins. The present study was performed to investigate the neuroprotective effects of CGP3466B and to determine a potential PCMT1/Mst1 neuronal anti-apoptotic pathway after TBI. Double immunofluorescence staining demonstrated that PCMT1 and Mst1 are co-located in neurons. Administration of CGP3466B improved neurological function, downregulated the ROS level and alleviated brain edema at 24 h after TBI. CGP3466B alleviates neuronal apoptosis by increasing PCMT1 expression and subsequently inhibiting MST1 activation, resulting in changing the expression levels of Bax, Bcl-2 and active-caspase3. The TUNEL staining results also support the anti-apoptosis effects of CGP3466B. The anti-apoptotic effects of CGP3466B were abolished by chelerythrine, an Mst1 activator, without changing PCMT1 levels. In conclusion, our findings suggest CGP3466B may have a promising therapeutic potential by modulating PCMT1/Mst1 signaling pathway after TBI injury.
Sigma-Aldrich
Anti-PCMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.