MilliporeSigma
  • The N-terminal region of centrosomal protein 290 (CEP290) restores vision in a zebrafish model of human blindness.
The N-terminal region of centrosomal protein 290 (CEP290) restores vision in a zebrafish model of human blindness.
Human molecular genetics (2011-01-25)
Lisa M Baye, Xiaobai Patrinostro, Svetha Swaminathan, John S Beck, Yan Zhang, Edwin M Stone, Val C Sheffield, Diane C Slusarski
ABSTRACT
The gene coding for centrosomal protein 290 (CEP290), a large multidomain protein, is the most frequently mutated gene underlying the non-syndromic blinding disorder Leber's congenital amaurosis (LCA). CEP290 has also been implicated in several cilia-related syndromic disorders including Meckel-Gruber syndrome, Joubert syndrome, Senor-Loken syndrome and Bardet-Biedl syndrome (BBS). In this study, we characterize the developmental and functional roles of cep290 in zebrafish. An antisense oligonucleotide [Morpholino (MO)], designed to generate an altered cep290 splice product that models the most common LCA mutation, was used for gene knockdown. We show that cep290 MO-injected embryos have reduced Kupffer's vesicle size and delays in melanosome transport, two phenotypes that are observed upon knockdown of bbs genes in zebrafish. Consistent with a role in cilia function, the cep290 MO-injected embryos exhibited a curved body axis. Patients with LCA caused by mutations in CEP290 have reduced visual perception, although they present with a fully laminated retina. Similarly, the histological examination of retinas from cep290 MO-injected zebrafish revealed no gross lamination defects, yet the embryos had a statistically significant reduction in visual function. Finally, we demonstrate that the vision impairment caused by the disruption of cep290 can be rescued by expressing only the N-terminal region of the human CEP290 protein. These data reveal that a specific region of the CEP290 protein is sufficient to restore visual function and this region may be a viable gene therapy target for LCA patients with mutations in CEP290.
Sigma-Aldrich
(−)-Epinephrine (+)-bitartrate salt, solid