MilliporeSigma
  • Endometrial sarcomas: an immunohistochemical and JAZF1 re-arrangement study in low-grade and undifferentiated tumors.
Endometrial sarcomas: an immunohistochemical and JAZF1 re-arrangement study in low-grade and undifferentiated tumors.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2012-08-25)
Kiran Jakate, Farshad Azimi, Rola H Ali, Cheng-Han Lee, Blaise A Clarke, Golnar Rasty, Patricia A Shaw, Nataliya Melnyk, David G Huntsman, Stephane Laframboise, Marjan Rouzbahman
ABSTRACT
The current World Health Organization classification divides endometrial sarcomas into low-grade endometrial stromal sarcoma and undifferentiated endometrial sarcoma. Recent studies suggest undifferentiated endometrial sarcoma is a heterogeneous group and a subgroup with uniform nuclei is more akin to low-grade endometrial stromal sarcoma in terms of morphologic, immunohistochemical and genetic features. We classified endometrial sarcomas treated at our institution from 1998 to 2011 into low-grade endometrial stromal sarcoma and undifferentiated endometrial sarcoma, the latter being further categorized into a group with either uniform or pleomorphic nuclei. Morphological features, immunoprofile and fluorescence in situ hybridization rearrangements of JAZF1 and PHF1 genes were correlated with tumor category and outcome. A total of 40 cases were evaluated comprising 23 low-grade endometrial stromal sarcomas, 10 undifferentiated endometrial sarcomas with nuclear uniformity and 7 undifferentiated endometrial sarcomas with nuclear pleomorphism. Low-grade endometrial stromal sarcomas were more often estrogen and progesterone receptor positive (83%) compared with undifferentiated endometrial sarcoma with nuclear uniformity (10%) or with nuclear pleomorphism (0%) (P<0.001). Positivity for p53 was restricted to undifferentiated endometrial sarcomas with more frequent expression in the group with nuclear pleomorphism (57%) than with nuclear uniformity (10%) (P=0.06). Ki-67 proliferation index in >10% of tumor cells more frequent in undifferentiated endometrial sarcoma than low-grade endometrial stromal sarcoma (P=<0.001). JAZF1 rearrangement was detected in 32% of low-grade endometrial stromal sarcomas and in none of the undifferentiated sarcomas. Rearrangement of PHF1 was found in two patients, one with JAZF1-PHF1 fusion. There were no significant differences in clinical behavior between undifferentiated endometrial sarcoma with nuclear uniformity versus nuclear pleomorphism. In conclusion, we found undifferentiated endometrial sarcoma subtypes and low-grade endometrial stromal sarcoma have distinct immunohistochemical and cytogentic profiles. Our data do not show any difference in clinical behavior between subgroups in undifferentiated sarcomas.