Skip to Content
MilliporeSigma
Zopiclone-N-oxide solution 100 μg/mL (Methanol with 1% 1 M HCl), ampule of 1 mL, certified reference material, Cerilliant®

Zopiclone-N-oxide solution

Zopiclone-N-oxide solution 100 μg/mL (Methanol with 1% 1 M HCl), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17H17ClN6O4
CAS Number:
Molecular Weight:
404.81

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing