Skip to Content
MilliporeSigma
Vildagliptin ≥98% (HPLC)

Vildagliptin

Vildagliptin ≥98% (HPLC)
Synonym(s):
Vildagliptin, (S)-1-(((1R,3R,5R,7S)-3-hydroxyadamantan-1-yl)glycyl)pyrrolidine-2-carbonitrile
Empirical Formula (Hill Notation):
C17H25N3O2
CAS Number:
Molecular Weight:
303.40

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing