![TSKgel® SuperSW mAb HTP Guard Column phase diol, L × I.D. 2 cm × 3 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/320/322/6155d20d-9ed4-440d-a447-06ea83bb0ded/640/6155d20d-9ed4-440d-a447-06ea83bb0ded.jpg)
TSKgel® SuperSW mAb HTP (4 µm) HPLC Column
![TSKgel® SuperSW mAb HTP Guard Column phase diol, L × I.D. 2 cm × 3 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/320/322/6155d20d-9ed4-440d-a447-06ea83bb0ded/640/6155d20d-9ed4-440d-a447-06ea83bb0ded.jpg)
Synonym(s):
TSKgel® Size Exclusion (SW-Type) HPLC Column, TSKgel® Size Exclusion (SW-Type) HPLC Guard Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2