![TSKgel® G2000PW HPLC Column phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 12 μm particle size](/deepweb/assets/sigmaaldrich/product/images/368/297/e1f95211-e3f8-42bd-bb9d-6d23e3e711fb/640/e1f95211-e3f8-42bd-bb9d-6d23e3e711fb.jpg)
TSKgel® G2000PW (12 µm) HPLC Columns
![TSKgel® G2000PW HPLC Column phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 12 μm particle size](/deepweb/assets/sigmaaldrich/product/images/368/297/e1f95211-e3f8-42bd-bb9d-6d23e3e711fb/640/e1f95211-e3f8-42bd-bb9d-6d23e3e711fb.jpg)
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2