
trans-2-Methyl-2-butenal

Synonym(s):
Tiglic aldehyde, (E)-2-Methylbut-2-enal, trans-2,3-Dimethylacrolein, trans-2-Methyl-2-butenal, Tiglinaldehyde
Linear Formula:
CH3CH=C(CH3)CHO
CAS Number:
Molecular Weight:
84.12
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1