Skip to Content
MilliporeSigma
Sodium octanoate-2,4,6,8-13C4 99 atom % 13C

Sodium octanoate-2,4,6,8-13C4

Sodium octanoate-2,4,6,8-13C4 99 atom % 13C
Synonym(s):
Octanoic acid-2,4,6,8-13C4 sodium salt
Linear Formula:
13CH3(CH213CH2)3CO2Na
CAS Number:
Molecular Weight:
170.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing