Skip to Content
MilliporeSigma
Sitostanol-5,6,22,23-d4 97 atom % D, 95% (CP)

Sitostanol-5,6,22,23-d4

Sitostanol-5,6,22,23-d4 97 atom % D, 95% (CP)
Empirical Formula (Hill Notation):
C29D4H48O
CAS Number:
Molecular Weight:
420.75

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing