Skip to Content
MilliporeSigma
Retinyl acetate synthetic, matrix dispersion, 475,000-650,000 USP units/g

Retinyl acetate

Retinyl acetate synthetic, matrix dispersion, 475,000-650,000 USP units/g
Synonym(s):
Retinol acetate, Vitamin A acetate, Retinyl acetate solution, Vitamin A acetate solution
Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique