Skip to Content
MilliporeSigma
Parthenolide phyproof® Reference Substance

Parthenolide

Parthenolide phyproof® Reference Substance
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3