Skip to Content
MilliporeSigma
Orlistat United States Pharmacopeia (USP) Reference Standard

Orlistat

Orlistat United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(−)-Tetrahydrolipstatin, N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-hexyl-4-oxo-2-oxetanyl]methyl]dodecyl ester, Ro-18-0647, Orlistat
Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique