N-Nitrosodiethylamine (NDEA)
Synonym(s):
N-Ethyl-N-nitrosoethanamine, N-Nitrosodiethylamine, Diethylnitrosamine
Linear Formula:
(C2H5)2NNO
CAS Number:
Molecular Weight:
102.14
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2