
Neobavaisoflavone

Synonym(s):
7-Hydroxy-3-(4-hydroxy-3-(3-methyl-2-buten-1-yl)phenyl)-4H-1-benzopyran-4-one
Empirical Formula (Hill Notation):
C20H18O4
CAS Number:
Molecular Weight:
322.35
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1