Skip to Content
MilliporeSigma
N-Acetyl-DL-cysteine-2,3-13C2,15N 98 atom % 15N, 99 atom % 13C, 98% (CP)

N-Acetyl-DL-cysteine-2,3-13C2,15N

N-Acetyl-DL-cysteine-2,3-13C2,15N 98 atom % 15N, 99 atom % 13C, 98% (CP)
Synonym(s):
NAC-13C2,15N
Empirical Formula (Hill Notation):
13C2C3H915NO3S
Molecular Weight:
166.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing