
N-Acetylbenzoquinoneimine

Synonym(s):
N-(4-Oxo-1-cyclohexa-2,5-dienylidene)acetamide, N-Acetyl-p-benzo-quinoneimine, NAPQI
Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1