
Mitomycin C

Synonym(s):
Mitomycin C
Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
Products
Product Category
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1