Skip to Content
MilliporeSigma
vibrant-m

MF-Millipore Membrane Filter, 0.45 µm pore size, gridded

vibrant-m
Synonym(s):
MCE Membrane Filter, 0.45 μm Pore Size

Products

Product Category
Brand
Color
Manufacturer
Diameter (mm)
Quality Segment
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing