
Methyldopate hydrochloride

Synonym(s):
Methyldopa ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C12H17NO4 · HCl
CAS Number:
Molecular Weight:
275.73
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing