Skip to Content
MilliporeSigma
Melamine-d6 98 atom % D, 99% (CP)

Melamine-d6

Melamine-d6 98 atom % D, 99% (CP)
Synonym(s):
1,3,5-Triazine-2,4,6-triamine-d6
Empirical Formula (Hill Notation):
C3D6N6
CAS Number:
Molecular Weight:
132.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing