![Macrophage Colony-Stimulating Factor from mouse M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/201/117/a1d2e41e-5dd5-4a73-8e70-c8d9573f9be8/640/a1d2e41e-5dd5-4a73-8e70-c8d9573f9be8.jpg)
Macrophage Colony-Stimulating Factor from mouse
![Macrophage Colony-Stimulating Factor from mouse M-CSF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/images/201/117/a1d2e41e-5dd5-4a73-8e70-c8d9573f9be8/640/a1d2e41e-5dd5-4a73-8e70-c8d9573f9be8.jpg)
Synonym(s):
CSF-1, M-CSF
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing