Skip to Content
MilliporeSigma
L-Tyrosine disodium salt hydrate ≥98% (HPLC), powder

L-Tyrosine disodium salt hydrate

L-Tyrosine disodium salt hydrate ≥98% (HPLC), powder
Synonym(s):
L-3-(4-Hydroxyphenyl)alanine disodium salt hydrate
Empirical Formula (Hill Notation):
C9H9NNa2O3 · xH2O
CAS Number:
Molecular Weight:
225.15 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Research Area
Quality Segment
Optical Isomer
Technique