Iron(III) chloride anhydrous
Synonym(s):
Iron(III) chloride
Empirical Formula (Hill Notation):
Cl3Fe
CAS Number:
Molecular Weight:
162.20
Products
Product Category
Brand
Formula Weight
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing