Skip to Content
MilliporeSigma
Guanosine 5′-[γ-thio]triphosphate tetralithium salt ≥90% (contains < 10% GDP, HPLC), powder

Guanosine 5′-[γ-thio]triphosphate tetralithium salt

Guanosine 5′-[γ-thio]triphosphate tetralithium salt ≥90% (contains < 10% GDP, HPLC), powder
Synonym(s):
GTPγS tetralithium salt, GTP-γ-S-Li4, Guanosine 5′-O-(3-thiotriphosphate) tetralithium salt
Empirical Formula (Hill Notation):
C10H12Li4N5O13P3S
CAS Number:
Molecular Weight:
562.98
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1