Skip to Content
MilliporeSigma
Gly-Pro p-nitroanilide p-toluenesulfonate salt ~99% (TLC)

Gly-Pro p-nitroanilide p-toluenesulfonate salt

Gly-Pro p-nitroanilide p-toluenesulfonate salt ~99% (TLC)
Empirical Formula (Hill Notation):
C13H16N4O4 · C7H8O3S
CAS Number:
Molecular Weight:
464.49

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing