
Glutaric dialdehyde solution

Synonym(s):
Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial
Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1