![Friedelin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/462/499/99f03a2c-6462-4a19-962e-3faf0624ded7/640/99f03a2c-6462-4a19-962e-3faf0624ded7.png)
Friedelin
![Friedelin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/462/499/99f03a2c-6462-4a19-962e-3faf0624ded7/640/99f03a2c-6462-4a19-962e-3faf0624ded7.png)
Synonym(s):
(4β,5β,8α,9β,10α,13α,14β)-5,9,13-Trimethyl-24,25,26-trinoroleanan-3-one, DA-friedooleanan-3-one, Friedelan-3-one
Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing