Skip to Content
MilliporeSigma
Fmoc-Pro-OH ≥90% (HPLC)

Fmoc-Pro-OH

Fmoc-Pro-OH ≥90% (HPLC)
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-proline, Fmoc-L-proline, Fmoc-Pro-OH
Empirical Formula (Hill Notation):
C20H19NO4
CAS Number:
Molecular Weight:
337.37

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing