Skip to Content
MilliporeSigma
Ethylene glycol diacrylate 90%, technical grade

Ethylene glycol diacrylate

Ethylene glycol diacrylate 90%, technical grade
Synonym(s):
Ethylene diacrylate, Ethylene glycol dipropenoate
Linear Formula:
(H2C=CHCO2CH2-)2
CAS Number:
Molecular Weight:
170.16
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing