Skip to Content
MilliporeSigma
D-Ribose 5-phosphate disodium salt dihydrate ≥99% (TLC)

D-Ribose 5-phosphate disodium salt dihydrate

D-Ribose 5-phosphate disodium salt dihydrate ≥99% (TLC)
Empirical Formula (Hill Notation):
C5H9Na2O8P · 2H2O
CAS Number:
Molecular Weight:
310.10

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing