
D-Luciferin potassium salt

Synonym(s):
(S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt, Firefly luciferin potassium salt
Empirical Formula (Hill Notation):
C11H7KN2O3S2
CAS Number:
Molecular Weight:
318.41
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1