Skip to Content
MilliporeSigma
Digoxigenin-3-O-methylcarbonyl-ε-aminocaproic acid-N-hydroxysuccinimide ester >90% (elementary analysis), pkg of 5 mg, powder, pH range 2-13

Digoxigenin-3-O-methylcarbonyl-ε-aminocaproic acid-N-hydroxysuccinimide ester

Digoxigenin-3-O-methylcarbonyl-ε-aminocaproic acid-N-hydroxysuccinimide ester >90% (elementary analysis), pkg of 5 mg, powder, pH range 2-13
Synonym(s):
Digoxigenin NHS-ester, ε-(Digoxigenin-3-0-acetamido)caproic acid N-hydroxysuccinimide ester, DIG NHS
Empirical Formula (Hill Notation):
C35H50N2O10
CAS Number:
Molecular Weight:
658.78

Products

Formula Weight
Manufacturer
pH Value
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing