
Copper(I) bromide dimethyl sulfide complex

Synonym(s):
Copper(I) bromide methyl sulfide complex, Cuprous bromide methyl sulfide complex
Linear Formula:
CuBr · CH3SCH3
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1