Skip to Content
MilliporeSigma
Chaetocin from Chaetomium minutum ≥95% (HPLC)

Chaetocin from Chaetomium minutum

Chaetocin from Chaetomium minutum ≥95% (HPLC)
Empirical Formula (Hill Notation):
C30H28N6O6S4
CAS Number:
Molecular Weight:
696.84

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing