
Cefquinome sulfate

Synonym(s):
(6R,7R)-7-[[(2Z)-2-(2-Amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-3-(5,6,7,8-tetrahydroquinolin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate sulfate, Cefquinome sulphate
Empirical Formula (Hill Notation):
C23H24N6O5S2 · H2SO4
CAS Number:
Molecular Weight:
626.68
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1